Methylene Blue hydrate, pure
Methylene Blue hydrate, pure
Methylene Blue hydrate, pure
Thermo Scientific Chemicals

Methylene Blue hydrate, pure

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 414250025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS122965-43-9
IUPAC Name7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC16H18ClN3S
InChI KeyCXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<22 %
Appearance (Color)Green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with TiCl3>=70 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only