Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-iodobenzoate, 98%

CAS: 619-44-3 | C8H7IO2 | 262.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 414460250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS619-44-3
IUPAC Namemethyl 4-iodobenzoate
Molecular FormulaC8H7IO2
InChI KeyDYUWQWMXZHDZOR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point113°C to 117°C
Appearance (Color)White to beige to light orange
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only