Methyl (R)-(+)-lactate, 98%
Methyl (R)-(+)-lactate, 98%
Methyl (R)-(+)-lactate, 98%
Thermo Scientific Chemicals

Methyl (R)-(+)-lactate, 98%

CAS: 17392-83-5 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 414490250
Price (USD)
210.65
Special offer
Online exclusive
Ends: 28-Dec-2026
248.00
Save 37.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
210.65
Special offer
Online exclusive
Ends: 28-Dec-2026
248.00
Save 37.35
Each
Chemical Identifiers
CAS17392-83-5
IUPAC Namemethyl 2-hydroxypropanoate
Molecular FormulaC4H8O3
InChI KeyLPEKGGXMPWTOCB-UHFFFAOYNA-N
SMILESCOC(=O)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4110 to 1.4150 (20°C, 589 nm)
Appearance (Form)Liquid
Optical rotation+6.5° to +10.5° (20°C, 589 nm) (neat)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only