(R)-(-)-1-Methyl-3-phenylpropylamine, 98%
(R)-(-)-1-Methyl-3-phenylpropylamine, 98%
(R)-(-)-1-Methyl-3-phenylpropylamine, 98%
Thermo Scientific Chemicals

(R)-(-)-1-Methyl-3-phenylpropylamine, 98%

CAS: 937-52-0 | C10H15N | 149.24 g/mol
Quantity:
1 g
5 g
Catalog number 414760050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS937-52-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Refractive index1.5130 to 1.5150 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only