3-Methylphthalic anhydride, 96%
3-Methylphthalic anhydride, 96%
3-Methylphthalic anhydride, 96%
Acros Organics

3-Methylphthalic anhydride, 96%

CAS: 4792-30-7 | C9H6O3 | 162.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
414780050
also known as 41478
5 gGlass bottle
Catalog number 414780050
also known as 41478
HSN 29173930
Price (INR)
18,148.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
18,148.00
Each
Chemical Identifiers
CAS4792-30-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige to light brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point114°C to 118°C
Titration with TBAH>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only