Methylsulfuric acid potassium salt, 97%
Methylsulfuric acid potassium salt, 97%
Methylsulfuric acid potassium salt, 97%
Thermo Scientific Chemicals

Methylsulfuric acid potassium salt, 97%

CAS: 562-54-9 | CH3KO4S | 150.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 414891000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS562-54-9
IUPAC Namepotassium methyl sulfate
Molecular FormulaCH3KO4S
InChI KeyWBGWGHYJIFOATF-UHFFFAOYSA-M
SMILES[K+].COS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Titration after Ion exchange96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only