Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Thermo Scientific™

Benchtop Dewar Flasks

Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
4150-10001 LCase of 4
4150-1000PK
also known as 4150-1000
1 LEach
4150-20002 LCase of 2
4150-2000PK
also known as 4150-2000
2 LEach
4150-40004 LCase of 2
4150-4000PK
also known as 4150-4000
4 LEach
4150-900010 LEach
Catalog number 4150-1000
Price (USD)
1,474.39
Online exclusive
1,720.43
Save 246.04
Case of 4
Capacity (Metric):
1 L
Unit Size:
Case of 4
Price (USD)
1,474.39
Online exclusive
1,720.43
Save 246.04
Case of 4

Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.

  • High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
  • Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
  • Can be used from -196°C to +100°C
  • Pour spout and bottom grip area for ease of removal of used liquids
  • Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifications
Capacity (English)33.814 fl oz.
Depth (English) Interior7.7 in.
Diameter (English) Inner Mouth3.8 in.
Diameter (Metric) Inner Mouth96mm
Height (English)9.9 in.
Height (Metric)251mm
No. per Case4
No. per Pack1
Capacity (Metric)1 L
Depth (Metric) Interior195 mm
Unit SizeCase of 4