Hypersil™ PREP HS Phenyl Bulk Media
Hypersil™ PREP HS Phenyl Bulk Media
Actual product may vary
Thermo Scientific™
Hypersil™ PREP HS Phenyl Bulk Media
Ensure optimal chromatography performance and cost savings with Hypersil PREP HS Phenyl bulk chromatography media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
41510-01000B1000 g
41510-00050B50 g
41510-00100B100 g
41510-00500B500 g
41510-05000B5000 g
41510-10000B10000 g
Catalog number 41510-01000B
Price (MXN)
-
Quantity:
1000 g
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil™ PREP HS Phenyl bulk chromatography media. Purchase bulk quantities of moderately hydrophobic, high surface area, phenyl-bonded media to repack preparative HPLC columns yourself without paying for new hardware.
Repack Columns Yourself with Bulk Media
Unique Selectivity
  • Enhanced selectivity for aromatic and moderately polar analytes.
  • Selectivity complementary to that provided by alkyl chain phases.
  • Moderate hydrophobicity compared to C18.
  • Phenyl (USP L11) selectivity - high surface area particle with 100Å pore size.
  • pH stable from 2 to 8.
  • Maximum temperature of 60°C.
Customer Specific Orders
  • Please contact your local sales representative for Customer Specific Orders.
Specifications
Carbon Load9%
Column TypeReversed Phase
EndcappedYes
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
PhaseReversed Phase
Pore Size100 Å
Product LinePrep LC
Quantity1000 g
Stationary PhasePhenyl (Ph)
Surface Area (Metric)350 m²/g
Temperature60°C
USP TypeL11
Particle Size10 μm
pH2 to 8
Unit SizeEach