Morpholine, 99+%, ACS reagent
Morpholine, 99+%, ACS reagent
Morpholine, 99+%, ACS reagent
Thermo Scientific Chemicals

Morpholine, 99+%, ACS reagent

CAS: 110-91-8 | C4H9NO | 87.12 g/mol
数量:
500 g
2.5 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 415160025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-91-8
IUPAC Namemorpholine
Molecular FormulaC4H9NO
InChI KeyYNAVUWVOSKDBBP-UHFFFAOYSA-N
SMILESC1COCCN1
さらに表示
Titration with HCl>=99.0 %
Color scale=<15 APHA
Infrared spectrumConforms
Boiling point126.0°C to 130.0°C
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only