Nickel phthalocyanine, 95%
Nickel phthalocyanine, 95%
Nickel phthalocyanine, 95%
Thermo Scientific Chemicals

Nickel phthalocyanine, 95%

CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 415600050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14055-02-8
IUPAC Namenickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular FormulaC32H16N8Ni
InChI KeyLVIYYTJTOKJJOC-UHFFFAOYSA-N
SMILES[Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Gravimetry>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only