Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 98+%, ACS reagent

CAS: 10101-97-0 | NiO46H2O | 262.85 g/mol
数量:
100 g
500 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 415610051
価格(JPY)
-
数量:
5 kg
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
さらに表示
Titration Complexometric98.0 to 102.0 %
Insoluble matter=<0.005 %
Nitrogen compounds (as N)=<0.002 %
Chloride (Cl)=<0.001 %
Calcium (Ca)=<0.005 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only