Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 98+%, ACS reagent

CAS: 10101-97-0 | NiO46H2O | 262.85 g/mol
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 415615000
Price (JPY)
-
Request A Quote
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric98.0 to 102.0 %
Insoluble matter=<0.005 %
Nitrogen compounds (as N)=<0.002 %
Chloride (Cl)=<0.001 %
Calcium (Ca)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only