Ninhydrin, ACS reagent
Ninhydrin, ACS reagent
Ninhydrin, ACS reagent
Thermo Scientific Chemicals

Ninhydrin, ACS reagent

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 415720100
Price (USD)
56.90
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
56.90
Each
Chemical Identifiers
CAS485-47-2
IUPAC Name2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O4
InChI KeyFEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILESOC1(O)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Crystals
Infrared spectrumConforms
IdentificationPasses test
Melting pointPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only