Nitrilotriacetic acid, 99%
Nitrilotriacetic acid, 99%
Nitrilotriacetic acid, 99%
Thermo Scientific Chemicals

Nitrilotriacetic acid, 99%

Nitrilotriacetic acid, CAS # 139-13-9, is a tricarboxylic acid compound used for chemical chelation and elution in protein studies and the purification of certain earth compounds. | CAS: 139-13-9 | C6H9NO6 | 191.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 415742500
Price (USD)
61.30
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
61.30
Each
Chemical Identifiers
CAS139-13-9
IUPAC Name2-[bis(carboxymethyl)amino]acetic acid
Molecular FormulaC6H9NO6
InChI KeyMGFYIUFZLHCRTH-UHFFFAOYSA-N
SMILESOC(=O)CN(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration excess NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Nitrilotriacetic acid (2,2',2-nitrilotriacetic acid) is a tricarboxylic acid used for chemical purification


Applications

  • A known mutagen, suitable for cell culture experiments of DNA damage
  • Nitrilotriacetic acid can be used for protein elution using the His-tag method and chemical chelation
  • Nitrilotriacetic acid is capable of chelating Ca2+, Co2+, Cu2+, and Fe3+
  • Intended for sensitive applications such as HPLC
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only