4-Nitrophthalic acid, 99%
4-Nitrophthalic acid, 99%
4-Nitrophthalic acid, 99%
Thermo Scientific Chemicals

4-Nitrophthalic acid, 99%

CAS: 610-27-5 | C8H5NO6 | 211.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 416120250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS610-27-5
IUPAC Name4-nitrobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H5NO6
InChI KeySLBQXWXKPNIVSQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point163°C to 167°C
Titration with TBAH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only