4-Nitrophthalonitrile, 99%
4-Nitrophthalonitrile, 99%
4-Nitrophthalonitrile, 99%
Thermo Scientific Chemicals

4-Nitrophthalonitrile, 99%

CAS: 31643-49-9 | C8H3N3O2 | 173.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 416150050
Price (EUR)
74,70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
74,70
Each
Chemical Identifiers
CAS31643-49-9
IUPAC Name4-nitrobenzene-1,2-dicarbonitrile
Molecular FormulaC8H3N3O2
InChI KeyNTZMSBAAHBICLE-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to light green to light grey
Melting point140°C to 144°C
HPLC>=98.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only