Olive oil, pure, refined
Thermo Scientific Chemicals

Olive oil, pure, refined

CAS: 8001-25-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 416530250
Price (USD)
22.60
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
22.60
Each
Chemical Identifiers
CAS8001-25-0
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4670 to 1.4710 (20°C, 589 nm)
Absorbance=<1.20 (1g/100ml cyclohexane) measured at the max.
Absorbancebetween 264nm and 276nm
Acid value=<0.3 mg KOH/g
Peroxide value=<10.0 meq O2/kg
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only