What does the unit TU/mg mean for enzyme activity versus the commonly used unit U/mg?
Papain activity is measured in terms of units of tyrosine release per mg (TU/mg), and this unit can be used interchangeably with the more general units per mg (U/mg).