Papain, for Biochemistry
Thermo Scientific Chemicals

Papain, for Biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
10g
100g
500g
Catalog number 416765000
Price (USD)
287.65
Online exclusive
319.00
Save 31.35
Each
Quantity:
500g
Request bulk or custom format
Price (USD)
287.65
Online exclusive
319.00
Save 31.35
Each
Chemical Identifiers
CAS9001-73-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
Assay>=1000 TU/mg (Enz.act.) accurancy:+/-5%
Loss on drying=<7.0 % (60°C, 4 hrs) (vacuum)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What does the unit TU/mg mean for enzyme activity versus the commonly used unit U/mg?

Papain activity is measured in terms of units of tyrosine release per mg (TU/mg), and this unit can be used interchangeably with the more general units per mg (U/mg).