Phenolphthalein, ACS reagent
Phenolphthalein, ACS reagent
Phenolphthalein, ACS reagent
Thermo Scientific Chemicals

Phenolphthalein, ACS reagent

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 417180026
Price (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Clarity of alcohol solutionPasses test
Visual transition intervalFrom pH 8.0 (colorless) to pH 10 (red)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only