Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
Thermo Scientific Chemicals

Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure

CAS: 62625-16-5 | C20H13K3O11S2 | 610.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 417220050
Price (USD)
178.65
Special offer
Online exclusive
Ends: 28-Dec-2026
210.00
Save 31.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 28-Dec-2026
210.00
Save 31.35
Each
Chemical Identifiers
CAS1080664-81-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only