Phosphomolybdic Acid hydrate, ACS Reagent
Phosphomolybdic Acid hydrate, ACS Reagent
Phosphomolybdic Acid hydrate, ACS Reagent
Thermo Scientific Chemicals

Phosphomolybdic Acid hydrate, ACS Reagent

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 417891000
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS51429-74-4
IUPAC Name12-Molybdophosphoric acid
Molecular FormulaH3Mo12O40P
Molecular Weight (g/mol)1825.40
Synonymphosphoric acid dodecamolybdenum hexatriacontaoxidandiide
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-orange
Appearance (Form)Adhering crystals
Heavy metals (as Pb)=<50 ppm
Insoluble matter=<0.01 %
Chloride (Cl)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only