Potassium hydrogen phthalate, 99+%
Potassium hydrogen phthalate, 99+%
Potassium hydrogen phthalate, 99+%
Thermo Scientific Chemicals

Potassium hydrogen phthalate, 99+%

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol
数量:
500 g
2.5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 417950025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
さらに表示
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only