Pinacol, 99%
Pinacol, 99%
Pinacol, 99%
Thermo Scientific Chemicals

Pinacol, 99%

Pinacol, 99%, C6H14O2, CAS Number-76-09-5, pinacone, pinacol, unii-527qe7i5co, tetramethylethylene glycol, 2,3-dimethyl-2,3-butanediol, 2,3-dimethyl-2,3-dihydroxybutane, 1,1,2,2-tetramethylethylene glycol, 2,3-butanediol, 2,3-dimethyl, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-butane-2,3-diol | CAS: 76-09-5 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 418002500
Price (EUR)
327,00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
327,00
Each
Chemical Identifiers
CAS76-09-5
IUPAC Name2,3-dimethylbutane-2,3-diol
Molecular FormulaC6H14O2
InChI KeyIVDFJHOHABJVEH-UHFFFAOYSA-N
SMILESCC(C)(O)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only