Potassium metabisulfite, 97%, extra pure
Potassium metabisulfite, 97%, extra pure
Potassium metabisulfite, 97%, extra pure
Thermo Scientific Chemicals

Potassium metabisulfite, 97%, extra pure

CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 418275000
Price (USD)
51.65
Online exclusive
56.70
Save 5.05
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Online exclusive
56.70
Save 5.05
Each
Chemical Identifiers
CAS16731-55-8
IUPAC Namedipotassium sulfinatosulfonate
Molecular FormulaK2O5S2
InChI KeyRWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Titration Iodimetric>=96.0 %
Heavy metals (as Pb)=<10 ppm
Selenium (Se)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only