Rhodamine B, 99+%, pure, laser grade
Rhodamine B, 99+%, pure, laser grade
Rhodamine B, 99+%, pure, laser grade
Thermo Scientific Chemicals

Rhodamine B, 99+%, pure, laser grade

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
Have Questions?
Catalog NumberQuantity
4190000101 g
Catalog number 419000010
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.60
Save 11.95
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.60
Save 11.95
Each
Chemical Identifiers
CAS81-88-9
IUPAC Name9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyPYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Optimum lasing588 nm (in Methanol)
UV Lambda max545 nm (in Methanol)
Molar absorption coefficient106000 l/(mol.cm) (at Lambda max)
Fluorescence max565 nm (in Methanol)
Appearance (Color)Green
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only