Rose Bengal, 85%, pure, certified
Rose Bengal, 85%, pure, certified
Rose Bengal, 85%, pure, certified
Thermo Scientific Chemicals

Rose Bengal, 85%, pure, certified

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 419151000
Price (USD)
156.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
156.00
Each
Chemical Identifiers
CAS632-69-9
IUPAC Namedisodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular FormulaC20H2Cl4I4Na2O5
InChI KeyKCQREHTWEUECQT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Absorption Ratio P-15/P+151.21 to 1.95
BSC CertificationPasses
Appearance (Color)Red-brown
Appearance (Form)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only