Diethyldithiocarbamic acid, silver salt, ACS reagent
Diethyldithiocarbamic acid, silver salt, ACS reagent
Diethyldithiocarbamic acid, silver salt, ACS reagent
Thermo Scientific Chemicals

Diethyldithiocarbamic acid, silver salt, ACS reagent

CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
数量:
10 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 419331000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1470-61-7
IUPAC Namesilver(1+) (diethylcarbamothioyl)sulfanide
Molecular FormulaC5H10AgNS2
InChI KeyNSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES[Ag+].CCN(CC)C([S-])=S
さらに表示
Appearance (Color)Yellow to green to khaki
Appearance (Form)Powder
Infrared spectrumConforms
Solubility(1 g/200 ml Pyridine) Clear bright yellow solution
Suitability>=0.10 Absorbance (for determination of Arsenic)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only