Thermo Scientific Chemicals

Silver nitrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 419365000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Silver nitrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 419365000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419365000
Price (USD)
Price: 1,424.00
Online price: 1,210.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7761-88-8
IUPAC Name
silver(1+) nitrate
Molecular Formula
AgNO3
InChI Key
SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES
[Ag+].[O-][N+]([O-])=O
Cadmium (Cd)
=<1 ppm
Nickel (Ni)
=<0.2 ppm
Palladium (Pd)
=<0.1 ppm
Zinc (Zn)
=<10 ppm
Chloride (Cl)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text