Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Thermo Scientific Chemicals

Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL

CAS: 5137-46-2 | C12H10Na | 177.202 g/mol
Have Questions?
Catalog NumberQuantity
419423000300 mL
Catalog number 419423000
Price (USD)
261.00
Each
Quantity:
300 mL
Request bulk or custom format
Price (USD)
261.00
Each
Chemical Identifiers
CAS5137-46-2
IUPAC Namesodium 1,1'-biphenyl hydride
Molecular FormulaC12H11Na
InChI KeySZJALDBKOTZIPO-UHFFFAOYSA-N
SMILES[H-].[Na+].C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
AppearanceMay partially crystallize
Titration with HClO415 to 25 % w/w
Appearance (Color)Dark green to blue-black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only