Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Catalog number 419530010
Price (USD)
390.65
Special offer
Online exclusive
Ends: 28-Dec-2026
459.00
Save 68.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
390.65
Special offer
Online exclusive
Ends: 28-Dec-2026
459.00
Save 68.35
Each
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<500 ppm
UVat 260 nm A: =<0.05
UVat 280 nm A: =<0.04
Solubility(3 % in water) Clear colorless
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only