Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Thermo Scientific Chemicals

Sodium polyanetholesulfonate

CAS: 55963-78-5 | (C10H11NaO4S)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 419631000
Price (USD)
393.65
Special offer
Online exclusive
Ends: 30-Jun-2026
463.00
Save 69.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
393.65
Special offer
Online exclusive
Ends: 30-Jun-2026
463.00
Save 69.35 (15%)
Each
Chemical Identifiers
CAS55963-78-5
Molecular Formula(C10H11NaO4S)n
MDL NumberMFCD00148427
SpecificationsSpecification SheetSpecification Sheet
pH5.0 to 7.0 (1 % in water)
Infrared spectrumConforms
Solubility(1% in water) Clear to hazy yellow solution,
Solubilityfree from visible impurities
Appearance (Color)White to yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only