LYVE1 Monoclonal Antibody (ALY7), eFluor™ 615, eBioscience™ - FAQs
LYVE1 Monoclonal Antibody (ALY7), eFluor™ 615, eBioscience™ - FAQs
View additional product information for LYVE1 Monoclonal Antibody (ALY7), eFluor™ 615, eBioscience™ - FAQs (42-0443-80, 42-0443-82)