Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs (42-4015-80, 42-4015-82)