Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 615, eBioscience™ - FAQs
Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 615, eBioscience™ - FAQs (42-4732-80)