FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 615, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 615, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 615, eBioscience™ - FAQs (42-5773-82)