GFAP Monoclonal Antibody (GA5), eFluor™ 615, eBioscience™ - FAQs
GFAP Monoclonal Antibody (GA5), eFluor™ 615, eBioscience™ - FAQs
View additional product information for GFAP Monoclonal Antibody (GA5), eFluor™ 615, eBioscience™ - FAQs (42-9892-80, 42-9892-82)