CD227 (Mucin 1) Monoclonal Antibody (SM3), eFluor™ 615, eBioscience™ - FAQs
CD227 (Mucin 1) Monoclonal Antibody (SM3), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD227 (Mucin 1) Monoclonal Antibody (SM3), eFluor™ 615, eBioscience™ - FAQs (42-9893-82, 42-9893-80)