Vimentin Monoclonal Antibody (V9), eFluor™ 615, eBioscience™ - FAQs
Vimentin Monoclonal Antibody (V9), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Vimentin Monoclonal Antibody (V9), eFluor™ 615, eBioscience™ - FAQs (42-9897-82)