3,3',4',5-Tetrachlorosalicylanilide, 97%
3,3',4',5-Tetrachlorosalicylanilide, 97%
3,3',4',5-Tetrachlorosalicylanilide, 97%
Thermo Scientific Chemicals

3,3',4',5-Tetrachlorosalicylanilide, 97%

CAS: 1154-59-2 | C13H7Cl4NO2 | 351.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 420230050
Price (EUR)
45,65
Online exclusive
50,90
Save 5,25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
45,65
Online exclusive
50,90
Save 5,25
Each
Chemical Identifiers
CAS1154-59-2
IUPAC Name3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Molecular FormulaC13H7Cl4NO2
InChI KeySJQBHPJLLIJASD-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only