Urea, 98.0-102.0%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Urea, 98.0-102.0%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets U.S.P, B.P., E.P, and J.P Specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
12 kg
85 lb.
50 kg
Número de catálogo 4203-08
Precio (MXN)
-
Cantidad:
85 lb.
This product is GMP Manufactured and meets U.S.P, B.P., E.P, and J.P Specifications.
Especificaciones
Punto de ebullición197°C (1013 hPa)
GradoMulti-Compendial
Peso molecular (g/mol)60.06
Porcentaje de pureza98.0-102.0%
Forma físicaCrystalline Powder
Cantidad85 lb.
Unit SizeEach