Tetrahexylammonium Bromide, 99%
Tetrahexylammonium Bromide, 99%
Tetrahexylammonium Bromide, 99%
Thermo Scientific Chemicals

Tetrahexylammonium Bromide, 99%

CAS: 4328-13-6 | C24H52BrN | 434.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 420400250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4328-13-6
IUPAC Nametetrahexylazanium bromide
Molecular FormulaC24H52BrN
InChI KeySYZCZDCAEVUSPM-UHFFFAOYSA-M
SMILES[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Appearance (Form)Powder or flakes
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only