Tetrakis(2-hydroxyethyl)ammonium bromide, 99%
Tetrakis(2-hydroxyethyl)ammonium bromide, 99%
Tetrakis(2-hydroxyethyl)ammonium bromide, 99%
Tetrakis(2-hydroxyethyl)ammonium bromide, 99%
Thermo Scientific Chemicals

Tetrakis(2-hydroxyethyl)ammonium bromide, 99%

CAS: 4328-04-5 | C8H20BrNO4 | 274.15 g/mol
製品番号(カタログ番号)数量パッケージ
42047025025 gGlass bottle
製品番号(カタログ番号) 420470250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4328-04-5
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only