N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
Thermo Scientific Chemicals

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%

CAS: 140-07-8 | C10H24N2O4 | 236.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 420480010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS140-07-8
IUPAC Name2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol
Molecular FormulaC10H24N2O4
InChI KeyBYACHAOCSIPLCM-UHFFFAOYSA-N
SMILESOCCN(CCO)CCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Titration with HClO4>=98.5 %
Refractive index1.4990 to 1.5070 (20°C, 589 nm)
Appearance (Form)Viscous liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only