Tetramethylammonium hydroxide, 25% in water
Tetramethylammonium hydroxide, 25% in water
Tetramethylammonium hydroxide, 25% in water
Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 25% in water

CAS: 75-59-2 | C4H13NO | 91.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 420520050
Price (USD)
603.65
Online Exclusive
670.00
Save 66.35 (10%)
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
603.65
Online Exclusive
670.00
Save 66.35 (10%)
Each
Chemical Identifiers
CAS75-59-2
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with HCl23 to 27 %
Refractive index1.3780 to 1.3840 (20°C, 589 nm)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only