Tetramethylammonium perchlorate, 98%
Tetramethylammonium perchlorate, 98%
Tetramethylammonium perchlorate, 98%
Thermo Scientific Chemicals

Tetramethylammonium perchlorate, 98%

CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 420560250
Price (EUR)
133,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
133,00
Each
Specifications
Assay Percent Range97.5% min. (on dry base)
CAS2537-36-2
Chemical Name or MaterialTetramethylammonium perchlorate
ColorWhite
EINECS Number219-805-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only