Tetrapropylammonium hydroxide, 25% in water
Tetrapropylammonium hydroxide, 25% in water
Tetrapropylammonium hydroxide, 25% in water
Thermo Scientific Chemicals

Tetrapropylammonium hydroxide, 25% in water

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 420735000
Price (USD)
453.65
Special offer
Online exclusive
Ends: 28-Dec-2026
533.00
Save 79.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
453.65
Special offer
Online exclusive
Ends: 28-Dec-2026
533.00
Save 79.35
Each
Chemical Identifiers
CAS4499-86-9
IUPAC Nametetrapropylazanium hydroxide
Molecular FormulaC12H29NO
InChI KeyLPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES[OH-].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Titration with HClO424 to 26 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only