Tin, ACS reagent, 30 mesh
Tin, ACS reagent, 30 mesh
Tin, ACS reagent, 30 mesh
Thermo Scientific Chemicals

Tin, ACS reagent, 30 mesh

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 420991000
Price (USD)
82.90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
82.90
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Appearance (Form)Metallic granular powder
Titration Complexometric>=99.5 %
Antimony (Sb)=<200 ppm
Arsenic (As)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only