o-Tolidine dihydrochloride, 99%
o-Tolidine dihydrochloride, 99%
o-Tolidine dihydrochloride, 99%
Thermo Scientific Chemicals

o-Tolidine dihydrochloride, 99%

CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 421130250
Price (EUR)
59,30
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
59,30
Each
Chemical Identifiers
CAS612-82-8
IUPAC Namedihydrogen 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2
InChI KeyLUKPNZHXJRJBAN-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to light grey
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 % (on dry substance)
Water=<5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only