o-Tolidine dihydrochloride, 99%
o-Tolidine dihydrochloride, 99%
o-Tolidine dihydrochloride, 99%
Thermo Scientific Chemicals

o-Tolidine dihydrochloride, 99%

CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
檢視全部
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 421135000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
化學識別
CAS612-82-8
IUPAC Namedihydrogen 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2
InChI KeyLUKPNZHXJRJBAN-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Light beige to light grey
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 % (on dry substance)
Water=<5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only